Anti-PDK3 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302064EA 698 EUR
ORIGTA302064
Anti-PDK3 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PDK3 (pyruvate dehydrogenase kinase, isozyme 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR