Anti-CD19 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302071EA 698 EUR
ORIGTA302071
Anti-CD19 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR