Anti-PDHX Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302074EA 638 EUR
ORIGTA302074
Anti-PDHX Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PDHX (pyruvate dehydrogenase complex, component X)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR