Anti-CTDSP1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302078EA 638 EUR
ORIGTA302078
Anti-CTDSP1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: CTDSP1 (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR