Anti-FGFR4 Mouse Monoclonal Antibody

Toimittaja: OriGene
ORIGTA302089EA 698 EUR
ORIGTA302089
Anti-FGFR4 Mouse Monoclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FGFR4 (fibroblast growth factor receptor 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR