Anti-CDX2 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302103EA 638 EUR
ORIGTA302103
Anti-CDX2 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: CDX2 (caudal type homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR