Anti-UBQLN1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302119EA 638 EUR
ORIGTA302119
Anti-UBQLN1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunofluorescence: 1:100; Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: UBQLN1 (ubiquilin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR