Anti-ALDH6A1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302121EA 638 EUR
ORIGTA302121
Anti-ALDH6A1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunochemistry: 1:10-50; Western Blot: 1:1000

Type: Primary
Antigen: ALDH6A1 (aldehyde dehydrogenase 6 family, member A1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR