Anti-CDH3 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302124EA 638 EUR
ORIGTA302124
Anti-CDH3 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: CDH3 (cadherin 3, type 1, P-cadherin (placental))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR