Anti-PARP6 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302166EA 579 EUR
ORIGTA302166
Anti-PARP6 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PARP6 (poly (ADP-ribose) polymerase family, member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR