Anti-STAT3 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302174EA 698 EUR
ORIGTA302174
Anti-STAT3 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:1000

Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR