Anti-EHMT1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302190EA 579 EUR
ORIGTA302190
Anti-EHMT1 Rabbit Polyclonal Antibody
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: EHMT1 (euchromatic histone-lysine N-methyltransferase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR