Anti-AK3 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302218EA 698 EUR
ORIGTA302218
Anti-AK3 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: AK3 (adenylate kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR