Anti-TRPM8 Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302225EA 638 EUR
ORIGTA302225
Anti-TRPM8 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: TRPM8 (transient receptor potential cation channel, subfamily M, member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR