Anti-PARG Rabbit Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302286EA 637 EUR
ORIGTA302286
Anti-PARG Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: PARG (poly (ADP-ribose) glycohydrolase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR