Anti-ADAM33 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302404EA 454 EUR
ORIGTA302404
Anti-ADAM33 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:64000 Western Blot: 01-03µg/ml

Type: Primary
Antigen: ADAM33 (ADAM metallopeptidase domain 33)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR