Anti-AKR1B10 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302407EA 454 EUR
ORIGTA302407
Anti-AKR1B10 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:8000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: AKR1B10 (aldo-keto reductase family 1, member B10 (aldose reductase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR