Anti-APPL1 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302413EA 500 EUR
ORIGTA302413
Anti-APPL1 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: APPL1 (adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR