Anti-ACTR1A Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302419EA 454 EUR
ORIGTA302419
Anti-ACTR1A Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: ACTR1A (ARP1 actin-related protein 1 homolog A, centractin alpha (yeast))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR