Anti-PPP2R5B Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302424EA 547 EUR
ORIGTA302424
Anti-PPP2R5B Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 05-2µg/ml

Type: Primary
Antigen: PPP2R5B (protein phosphatase 2, regulatory subunit B', beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR