Anti-BHMT Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302433EA 547 EUR
ORIGTA302433
Anti-BHMT Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR