Anti-CASP4 Goat polyclonal antibody

Toimittaja: OriGene
ORIGTA302442EA 547 EUR
ORIGTA302442
Anti-CASP4 Goat polyclonal antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-2µg/ml

Type: Primary
Antigen: CASP4 (caspase 4, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR