Anti-CTNNA1 Goat polyclonal antibody

Toimittaja: OriGene
ORIGTA302443EA 454 EUR
ORIGTA302443
Anti-CTNNA1 Goat polyclonal antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: CTNNA1 (catenin (cadherin-associated protein), alpha 1, 102kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR