Anti-CORO1A Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302455EA 454 EUR
ORIGTA302455
Anti-CORO1A Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 025-1µg/ml

Type: Primary
Antigen: CORO1A (coronin, actin binding protein, 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR