Anti-CREM Goat polyclonal antibody

Toimittaja: OriGene
ORIGTA302456EA 547 EUR
ORIGTA302456
Anti-CREM Goat polyclonal antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: CREM (cAMP responsive element modulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR