Anti-CSRP2 Goat polyclonal antibody

Toimittaja: OriGene
ORIGTA302457EA 547 EUR
ORIGTA302457
Anti-CSRP2 Goat polyclonal antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:64000 Western Blot: 01-03µg/ml

Type: Primary
Antigen: CSRP2 (cysteine and glycine-rich protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR