Anti-CSNK1E Goat polyclonal antibody

Toimittaja: OriGene
ORIGTA302458EA 500 EUR
ORIGTA302458
Anti-CSNK1E Goat polyclonal antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: CSNK1E (casein kinase 1, epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR