Anti-DKK4 Goat polyclonal antibody

Toimittaja: OriGene
ORIGTA302466EA 550 EUR
ORIGTA302466
Anti-DKK4 Goat polyclonal antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: DKK4 (dickkopf WNT signaling pathway inhibitor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR