Anti-ELMO1 Goat polyclonal antibody

Toimittaja: OriGene
ORIGTA302474EA 454 EUR
ORIGTA302474
Anti-ELMO1 Goat polyclonal antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1-2µg/ml

Type: Primary
Antigen: ELMO1 (engulfment and cell motility 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR