Anti-PTGER4 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302476EA 547 EUR
ORIGTA302476
Anti-PTGER4 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: PTGER4 (prostaglandin E receptor 4 (subtype EP4))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR