Anti-ESRRG Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302478EA 454 EUR
ORIGTA302478
Anti-ESRRG Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 02-06µg/ml

Type: Primary
Antigen: ESRRG (estrogen-related receptor gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR