Anti-GCNT3 Goat polyclonal antibody

Toimittaja: OriGene
ORIGTA302496EA 454 EUR
ORIGTA302496
Anti-GCNT3 Goat polyclonal antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:2000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: GCNT3 (glucosaminyl (N-acetyl) transferase 3, mucin type)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR