Anti-GRAP2 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302501EA 454 EUR
ORIGTA302501
Anti-GRAP2 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 02-1µg/ml

Type: Primary
Antigen: GRAP2 (GRB2-related adaptor protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR