Anti-GRB7 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302502EA 547 EUR
ORIGTA302502
Anti-GRB7 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: GRB7 (growth factor receptor-bound protein 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR