Anti-ZDHHC17 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302507EA 547 EUR
ORIGTA302507
Anti-ZDHHC17 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: ZDHHC17 (zinc finger, DHHC-type containing 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR