Anti-IRF6 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302518EA 454 EUR
ORIGTA302518
Anti-IRF6 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1/32000; Western Blot: 02-1µg/ml

Type: Primary
Antigen: IRF6 (interferon regulatory factor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR