Anti-KLF15 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302522EA 550 EUR
ORIGTA302522
Anti-KLF15 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-10µg/ml

Type: Primary
Antigen: KLF15 (Kruppel-like factor 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR