Anti-KLK1 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302523EA 547 EUR
ORIGTA302523
Anti-KLK1 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:2000

Type: Primary
Antigen: KLK1 (kallikrein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR