Anti-LDHC Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302529EA 547 EUR
ORIGTA302529
Anti-LDHC Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 003-01µg/ml; Immunochemistry: 1?g/ml

Type: Primary
Antigen: LDHC (lactate dehydrogenase C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR