Anti-MAN2A1 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302533EA 547 EUR
ORIGTA302533
Anti-MAN2A1 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: MAN2A1 (mannosidase, alpha, class 2A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR