Anti-PCGF2 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302537EA 547 EUR
ORIGTA302537
Anti-PCGF2 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-2µg/ml

Type: Primary
Antigen: PCGF2 (polycomb group ring finger 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR