Anti-MEPCE Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302538EA 456 EUR
ORIGTA302538
Anti-MEPCE Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1-3?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: MEPCE (methylphosphate capping enzyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR