Anti-MYB Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302544EA 547 EUR
ORIGTA302544
Anti-MYB Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:1000; Western Blot: 0.5-2 ?g/ml; Immunochemistry: 3-6 ?g/ml

Type: Primary
Antigen: MYB (v-myb avian myeloblastosis viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR