Anti-BLOC1S6 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302559EA 500 EUR
ORIGTA302559
Anti-BLOC1S6 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: BLOC1S6 (biogenesis of lysosomal organelles complex-1, subunit 6, pallidin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR