Anti-PDE4D Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302564EA 500 EUR
ORIGTA302564
Anti-PDE4D Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml: 1-3 µg/ml

Type: Primary
Antigen: PDE4D (phosphodiesterase 4D, cAMP-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR