Anti-PDE5A Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302566EA 454 EUR
ORIGTA302566
Anti-PDE5A Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-2µg/ml

Type: Primary
Antigen: PDE5A (phosphodiesterase 5A, cGMP-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR