Anti-PSMF1 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302572EA 456 EUR
ORIGTA302572
Anti-PSMF1 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1/32000 Western Blot: 05-2µg/ml

Type: Primary
Antigen: PSMF1 (proteasome (prosome, macropain) inhibitor subunit 1 (PI31))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR