Anti-PIN1 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302573EA 454 EUR
ORIGTA302573
Anti-PIN1 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:128000; Western Blot: 02-06µg/ml

Type: Primary
Antigen: PIN1 (peptidylprolyl cis/trans isomerase, NIMA-interacting 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR