Anti-PLA2G1B Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302575EA 547 EUR
ORIGTA302575
Anti-PLA2G1B Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:64000; Western Blot: 001µg/ml

Type: Primary
Antigen: PLA2G1B (phospholipase A2, group IB (pancreas))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR