Anti-PNPLA2 Goat Polyclonal Antibody

Toimittaja: OriGene
ORIGTA302576EA 547 EUR
ORIGTA302576
Anti-PNPLA2 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:8000; Western Blot: 02-06µg/ml

Type: Primary
Antigen: PNPLA2 (patatin-like phospholipase domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR